Learn More
Medchemexpress LLC Enoblituzumab (Synonyms: MGA271) 10mg
SDP
Supplier:  Medchemexpress LLC HYP996610MG
Encompass_Preferred
Enoblituzumab (MGA271) is a humanized IgG1κ monoclonal antibody recognizing human B7-H3 protein, a member of the B7 family of immune regulators.
Catalog No. NC3401815