Learn More
Cayman Chemical Enobosarm-d4 500ug
SDP
Supplier:  Cayman Chemical 34687500 UG
Encompass_Preferred
An internal standard for the quantification of enobosarm by GC- or LC-MS
Catalog No. 50-291-0289