Learn More
Medchemexpress LLC Enocyanin 10g | 11029-12-2 | 10 G

Supplier: Medchemexpress LLC HY11433610G
Enocyanin is an anthocyanin pigment extracted from grape skins, supplied as a powder for biochemical research and enzyme inhibition studies.
- Appearance: solid, brown to reddish brown.
- Biological activity: inhibits leucine aminopeptidase, acid phosphatase, γ-glutamyl transpeptidase, and esterase activity.
- Storage: powder store at -20°C for long-term (3 years) or 4°C for shorter term (2 years).
- Available pack sizes range from 100 mg to 50 g, including a 10 G pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.