Learn More
Apexbio Technology LLC Enoxaparin 5mg
SDP
Supplier:  Apexbio Technology LLC BA22665
Encompass_Preferred
Enoxaparin (CAS 679809-58-6) is a low molecular weight heparin derivative that acts by potentiating the inhibitory activity of antithrombin III leading to the inhibition of coagulation factors such as thrombin (factor IIa) Through this mechanism Enoxaparin demonstrates anticoagulant properties Experimental studies indicate that it confers neuroprotection in rat hippocampus following traumatic brain injury via antioxidative and anti-inflammatory pathways Enoxaparin is widely used in research investigating deep vein thrombosis pulmonary embolism traumatic brain injury and thrombotic complications associated with COVID-19
Catalog No. 50-000-89378