Learn More
Medchemexpress LLC Enoxaparin | 679809-58-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109509100MG
Encompass_Preferred
Enoxaparin (PK 10169) is a low-molecular-weight heparin (LMWH) derivative that acts as an anticoagulant by inhibiting factor Xa and thrombin IIa through antithrombin III. It also demonstrates antioxidant and anti-inflammatory properties, protecting the rat hippocampus against traumatic brain injury (TBI). Enoxaparin is utilized in research for deep vein thrombosis (DVT), pulmonary embolism, TBI, and COVID-19.
  • Exerts anticoagulant activity through antithrombin III, inhibiting factor Xa and thrombin IIa.
  • Protects the rat hippocampus against TBI via antioxidant and anti-inflammatory properties.
  • Can be used for research on deep vein thrombosis (DVT), pulmonary embolism, TBI, and COVID-19.
  • In vitro, it enhances AAT (alpha-1-antitrypsin) inhibition of TMPRSS2 activity and HCoV-229E infection of hAEc.
  • In vivo, it reduces oxidative damage, inflammation, and astrocytosis following TBI in rats.
Catalog No. 50-003-45377