Learn More
Medchemexpress LLC Enoxaparin sodium | 679809-58-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10950950MG
Encompass_Preferred
Enoxaparin (PK 10169) is a low-molecular-weight heparin (LMWH) derivative that acts as an anticoagulant by inhibiting factor Xa and thrombin IIa through antithrombin III. It also possesses antioxidant and anti-inflammatory properties, protecting the rat hippocampus against traumatic brain injury (TBI). Enoxaparin is used in research for conditions such as deep vein thrombosis (DVT), pulmonary embolism, TBI, and COVID-19.
  • Low-molecular-weight heparin (LMWH) derivative.
  • Exerts anticoagulant activity.
  • Inhibits factor Xa and thrombin IIa.
  • Protects against TBI via antioxidant and anti-inflammatory properties.
  • Potential research applications include deep vein thrombosis (DVT), pulmonary embolism, TBI, and COVID-19.
Catalog No. 50-003-44701