Learn More
Selleck Chemical LLC Enoxaparin sodium S5415-25mg
SDP
Supplier:  Selleck Chemical LLC S541525MG
Encompass_Preferred
Enoxaparin sodium(PK 10169) is a low-molecular-weight heparin used to prevent and treat deep vein thrombosis or pulmonary embolism It binds to and accelerates the activity of antithrombin III
Catalog No. 50-310-9938