Learn More
Medchemexpress LLC HY-129490 5mg Medchemexpress, Enpp-1-IN-1 CAS:2289728-58-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1294905MG
Encompass_Preferred
Medchemexpress, HY-129490 5mg Enpp-1-IN-1 CAS:2289728-58-9 Enpp-1-IN-1 is a Ectonucleotide pyrophosphatase-phosphodiesterase 1 (enpp-1) inhibitor extracted from patent WO2019046778, Example 55. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9808