Learn More
Medchemexpress LLC Enpp-1-IN-10 | 2631704-38-4 | 98.64% | C13H12N6OS | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14325410MM/1ML
Encompass_Preferred
Enpp-1-IN-10 is a potent Ecto-nucleotide pyrophosphatase/phosphodiesterases 1 (ENPP1) inhibitor with a Ki value of 3.866 μM. This compound can be used for researching anticancer applications.
  • Potent ENPP1 inhibitor
  • Molecular weight: 300.34
  • Research use only
Catalog No. 50-003-55535