Learn More
Medchemexpress LLC Enpp/Carbonic anhydrase-in-1 | 2883495-35-8 | C₂₃H₂₅NOS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151916100MG
Encompass_Preferred
Enpp/Carbonic anhydrase-IN-1 (compound 1e) is a potent inhibitor of both Enpp and carbonic anhydrase. This compound demonstrates antiproliferative activity against cancer cells while showing low cytotoxicity towards normal cells. It is also known to induce apoptosis.
  • Potent inhibitor of Enpp and carbonic anhydrase
  • Exhibits specific IC₅₀ values for NPP1, NPP2, NPP3, CA-II, and CA-IX
  • Demonstrates antiproliferative activity against cancer cells
  • Shows low cytotoxicity against normal cells
  • Induces apoptosis
Catalog No. 50-003-74148