Learn More
Medchemexpress LLC Enpp/Carbonic anhydrase-IN-1 | 2883495-35-8 | C23H25NO4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1519165MG
Encompass_Preferred
Enpp/Carbonic anhydrase-IN-1 (compound 1e) is a potent Enpp and carbonic anhydrase inhibitor with IC50s of 1.36, 1.35, 3.00, 0.88, and 1.02 μM for NPP1, NPP2, NPP3, CA-II, and CA-IX respectively. It exhibits antiproliferative activity against cancer cells and low cytotoxicity against normal cells, and it induces Apoptosis.
  • Potent Enpp and carbonic anhydrase inhibitor.
  • Exhibits antiproliferative activity against cancer cells.
  • Low cytotoxicity against normal cells.
  • Induces apoptosis.
Catalog No. 50-003-73856