Learn More
Medchemexpress LLC Enpp/Carbonic anhydrase-IN-1 | 2883495-35-8 | C23H25NO4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15191650MG
Encompass_Preferred
Enpp/Carbonic anhydrase-IN-1 is a potent Enpp and carbonic anhydrase inhibitor with antiproliferative activity for cancer cells. It demonstrates low cytotoxicity against normal cells and induces apoptosis, making it suitable for research applications.
  • Potent Enpp and carbonic anhydrase inhibitor
  • Shows antiproliferative activity for cancer cells
  • Low cytotoxicity against normal cells
  • Induces apoptosis
Catalog No. 50-003-72774