Learn More
Cayman Chemical ENPP1 InhIbItr 43 5mg
SDP
Supplier:  Cayman Chemical 368365 MG
Encompass_Preferred
An ENPP1 inhibitor (Ki = 41 nM); inhibits tumor growth in an LLC1 murine lung cancer model when administered orally or intravenously at doses of 10 and 100 mg/kg, respectively
Catalog No. 50-290-9377