Learn More
Cayman Chemical ENPP3 InhIbItr 1mg
SDP
Supplier:  Cayman Chemical 377601 MG
Encompass_Preferred
An ENPP3 inhibitor (Ki = 53.7 nM); selective for ENPP3 over ENPP1, ENPP2, TNAP, CD73, NTPDase 1, NTPDase 2, and NTPDase 3; inhibits CAII and CAIX (Kis = 74.7 and 20.3 nM, respectively)
Catalog No. 50-291-7805