Learn More
Cayman Chemical ENPP3 InhIbItr 4g 5mg
SDP
Supplier:  Cayman Chemical 376475 MG
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.45 and 0.19 µM, respectively)
Catalog No. 50-291-5822