Learn More
Cayman Chemical ENPP3 InhIbItr 4t 25mg
SDP
Supplier:  Cayman Chemical 3764625 MG
Encompass_Preferred
An ENPP3 inhibitor (IC50 = 0.15 µM); selective for ENPP3 over ENPP1 (IC50 = 41.4 µM)
Catalog No. 50-291-5768