Learn More
Cayman Chemical EnrofloxacIn-d5 10mg
SDP
Supplier:  Cayman Chemical 3380910 MG
Encompass_Preferred
An internal standard for the quantification of enrofloxacin by GC- or LC-MS
Catalog No. 50-294-7436