Learn More
Medchemexpress LLC Enrofloxacin (monohydrochloride) | 93106-59-3 | 99.7% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0502A5G
Encompass_Preferred
Enrofloxacin monohydrochloride is an effective antibiotic that exhibits an MIC90 of 0.312 μg/mL against *Mycoplasma bovis*. It also demonstrates inhibitory activity against vaccinia virus (VV) and is intended for research use.
  • Effective antibiotic
  • Exhibits MIC90 of 0.312 μg/mL for Mycoplasma bovis
  • Shows inhibitory activity against vaccinia virus (VV)
  • High purity of 99.73%
  • Recommended storage at 4°C, sealed, away from moisture
Catalog No. 50-004-05272