Learn More
Medchemexpress LLC Enrofloxacin (Standard) | 93106-60-6 | 99.9% | 250 MG

Supplier: Medchemexpress LLC 250MGHYB0502R
Enrofloxacin (Standard) is presented as the analytical standard for Enrofloxacin, a potent antibiotic with an MIC90 of 0.312 μg/mL against *Mycoplasma bovis*. This product is specifically designed for research and analytical applications, serving as a reference standard in assays. It is commonly employed in qualitative, quantitative, and methodological research experiments utilizing techniques such as HPLC, GC, and MS.
- Synonyms: BAY Vp 2674 (Standard); PD160788 (Standard)
- CAS No.: 93106-60-6
- Purity: 99.94%
- Molecular weight: 359.39
- Formula: C₁₉H₂₂FN₃O₃
- Appearance: Solid
- Color: Off-white to light yellow
- Application: Used as an analytical standard in HPLC, GC, and MS for qualitative, quantitative, and methodological research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.