Learn More
Selleck Chemical LLC Ensartinib dihydrochloride S2934-5mg
SDP
Supplier:  Selleck Chemical LLC S29345MG
Encompass_Preferred
Ensartinib dihydrochloride is a potent new-generation ALK inhibitor with high activity against a broad range of known crizotinib-resistant ALK mutations and CNS metastases It potently inhibits both wild-type ALK and ALK variants (F1174 C1156Y L1196M S1206R T1151 and G1202R mutants) with in vitro IC50s of 4 nM
Catalog No. 50-311-2491