Learn More
TARGETMOL CHEMICALS INC Ensifentrine 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2537610MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ensifentrine (Ensifentrinum) is a PDE3/4 inhibitor, although its affinity for PDE3(IC50 0.4 nM) is 3,440 times higher than that for PDE4(IC50 1479 nM), that is under clinical development for the treatment of asthma and COPD and, potentially, cystic fibrosis. Purity 98.45%
Catalog No. 50-287-7535