Learn More
Medchemexpress LLC Ensitrelvir | 2647530-73-0 | >99.6% | C22H17ClF3N9O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14321610MM/1ML
Encompass_Preferred
Ensitrelvir (S-217622) is the first orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor. It is intended for research use only.
  • Orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor.
  • Inhibits intrapulmonary replication of SARS-CoV-2 in mice.
  • Shows antiviral activity against SARS-CoV-2 in Vero E6 cells.
Catalog No. 50-003-54980