Learn More
Medchemexpress LLC Ensitrelvir | 2647530-73-0 | 99.6% | C22H17ClF3N9O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY143216
Encompass_Preferred
Ensitrelvir (S-217622) is the first orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor (IC50=13 nM). It is for research use only.
  • Orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor.
  • Inhibits intrapulmonary replication of SARS-CoV-2 in mice dose-dependently.
  • Shows EC50 values of approximately 0.4 μM for both wild-type virus and Alpha, Beta, Gamma, and Delta variants of SARS-CoV-2 in a cytopathic effect (cpe)-inhibition assay in VeroE6/TMPRSS2 cells.
  • EC50 values for SARS-CoV and MERS-CoV were 0.21 and 1.4 μM, respectively.
Catalog No. 50-003-56850