Learn More
Medchemexpress LLC Ensitrelvir | 2647530-73-0 | MFCD34604124 | 99.6% | C22H17ClF3N9O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14321610MG
Encompass_Preferred
Ensitrelvir is an orally active, non-covalent, non-peptidic inhibitor of the SARS-CoV-2 3CL (main) protease intended for research use; it shows potent biochemical activity and is supplied as the free base solid with fumarate salt forms available.
  • Potent 3CL protease inhibition (IC50 = 13 nM).
  • Orally active, non-covalent, non-peptidic mechanism for antiviral research.
  • High reported purity (99.6%) for reproducible results.
  • Available as a solid free base; fumarate salt also available.
  • Suitable for in vitro antiviral and protease assays; for research use only.
  • Stable under recommended storage (powder: -20°C long term; in solvent: -80°C short term).
Catalog No. 50-002-61302