Learn More
Medchemexpress LLC Ensitrelvir fumarate | 2757470-18-9 | 99.99% | C26H21ClF3N9O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY143216A100MG
Encompass_Preferred
Ensitrelvir fumarate (S-217622) is the first orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor with an IC50 of 13 nM. It demonstrates EC50 values of approximately 0.4 μM against wild-type SARS-CoV-2 and its Alpha, Beta, Gamma, and Delta variants in cytopathic effect (cpe)-inhibition assays. This compound also dose-dependently inhibits the intrapulmonary replication of SARS-CoV-2 in mice and shows activity against SARS-CoV and MERS-CoV.
  • Orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor.
  • Inhibits wild-type SARS-CoV-2 and Alpha, Beta, Gamma, and Delta variants.
  • Inhibits intrapulmonary replication of SARS-CoV-2 in mice.
  • Exhibits activity against SARS-CoV and MERS-CoV.
Catalog No. 50-003-54473