Learn More
Medchemexpress LLC Ent-4-(6-azaspiro[2.5]octan-6-yl)-2-oxo-8-propyl-1,2,5,6,7,8-hexahydro-1,6-naphthyridine-3-carbonitr | 3036022-06-4 | 99.0% | 362.90 g/mol | C19H27ClN4O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15210610MG
Encompass_Preferred
BAY-7081 is a cyanopyridone-based chemical probe that potently and selectively inhibits phosphodiesterase 9A (PDE9A). It is orally active and soluble, with an in vitro IC50 of 15 nM, and is used in biochemical and cellular studies to probe PDE9A function and signaling.
  • Potent PDE9A inhibition (IC50 = 15 nM).
  • Selective for PDE9A over related phosphodiesterases.
  • Orally active and soluble, suitable for in vivo or in vitro studies.
  • High purity (≈99.0%) for reliable experimental results.
  • Provided as a purified small-molecule solid for biochemical assays.
  • Useful as a chemical probe to study PDE9A-mediated pathways.
Catalog No. 50-002-86255