Learn More
Medchemexpress LLC Entacapone acid | 160391-70-8 | 98.9% | 250.16 g/mol | C10H6N2O6 | 1MG

Supplier: Medchemexpress LLC HY1253501MG
Entacapone acid (Tyrphostin AG1290) is a research-grade tyrosine kinase inhibitor (CAS 160391-70-8) used as a biochemical tool to study kinase activity and hepatic protein synthesis. It is supplied as a purified powder with defined storage conditions for stability and long-term use.
- High purity: 98.9%
- Molecular weight: 250.16 g/mol
- Chemical formula: C10H6N2O6
- Reported biological activity: reduces hepatic protein synthesis in vivo
- Available in small research pack sizes, including 1 mg
- Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.