Learn More
Medchemexpress LLC Entacapone-d10 | 1185241-19-3 | 98.6% | C14H5D10N3O5 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY14280S500UG
Encompass_Preferred
Entacapone-d10 is a deuterium-labeled Entacapone. Entacapone is a potent, reversible, peripherally acting, and orally active catechol-O-methyltransferase (COMT) inhibitor. It inhibits COMT from rat brain, erythrocytes, and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. It is selective for COMT over other catecholamine metabolizing enzymes like MAO-A, MAO-B, phenolsulphotransferase M (PST-M), and PST-P (IC50s>50 μM). Entacapone also inhibits FTO demethylation with an IC50 of 3.5 μM.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled compound for drug development quantitation.
  • Potentially affects pharmacokinetic and metabolic profiles of drugs.
  • Useful for Parkinson's disease research.
  • Applicable for metabolic disorder research.
Catalog No. 50-003-55689