Learn More
ATCC Klebsiella aerogenes Tindall et al.
Supplier:  ATCC 13048/DEL
Encompass
Type strain. Used in assays and bacterial resistance testing, and as a quality control strain for BD and bioMerieux Vitek products. This strain is recommended by ATCC for use in the tests described in ASTM Standard Test Method D4783-89 where only the taxon is specified.
Catalog No. 50-189-135FP