Learn More
Cayman Chemical Enterocn 5mg
SDP
Supplier:  Cayman Chemical 205925 MG
Encompass_Preferred
An antibiotic that has static activities against a broad range of Gram-positive and Gram-negative bacteria but has no activity against fungi, including yeasts
Catalog No. 50-298-5422