Learn More
ATCC Enterococcus casseliflavus; Strain 9199
Supplier:  ATCC 700327
Encompass
Enterococcus casseliflavus strain 9199 is a whole-genome sequenced bacterium that can be used as a quality control for the VITEK 2 ID-GP card.
Catalog No. 50-238-3887