Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC 700802
Encompass
Enterococcus faecalis strain V583 was isolated in St. Louis, Missouri, from the blood of a patient. This whole-genome sequenced bacterial strain is resistant to gentamicin and vancomycin and the presence of the vanB gene was confirmed by PCR. The culture can be used in antimicrobial resistance research.
Catalog No. 50-238-3709