Learn More
ATCC Enterococcus faecium
Supplier:  ATCC BAA2320
Encompass
This strain of Enterococcus faecium was isolated from human feces. This whole-genome sequenced bacterium is multidrug resistant and was confirmed by PCR to carry the vanA gene. It can be used in antimicrobial resistance research.
Catalog No. 50-238-4467
 Restricted Item