Learn More
ATCC Enterococcus faecium; Strain AGR 15
Supplier:  ATCC BAA2127
Encompass
Enterococcus faecium strain AGR15 is a drug-resistant bacterium that was collected from piglet feces in Kansas. It is resistant to erythromycin and tetracycline, and no vancomycin resistance genes were detected by PCR. This bacterium has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-3721