Learn More
Medchemexpress LLC Enteropeptidase | 9014-74-8 | 500 U
SDP
Supplier:  Medchemexpress LLC HYP2990500U
Encompass_Preferred
Enteropeptidase (TMPRSS15) is a type II transmembrane serine protease that acts as a physiological activator of trypsinogen. It is primarily associated with the brush border membrane (BBM) of enterocytes in the upper small intestine and plays a crucial role in digestion.
  • Type II transmembrane serine protease
  • Activator of trypsinogen
  • Involved in digestion
  • For research use only
Catalog No. 50-004-00163