Learn More
ATCC Enterovirus D68;Strain: US/KY/14-18953
Supplier:  ATCC VR1825
Encompass
Enterovirus D68 strain US/KY/14-18953 is propagated in RD cells (ATCC CCL-136). This strain was isolated from a nasopharyngeal swab from a patient with respiratory illness in Kentucky in 2014 and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory disease research.
Catalog No. 50-238-5052