Learn More
Medchemexpress LLC HY-12163 200mg , Entinostat MS-275;SNDX-275 CAS:209783-80-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12163200MG
Encompass_Preferred
Medchemexpress, HY-12163 200mg Entinostat MS-275;SNDX-275 CAS:209783-80-2 Formula:C21H20N4O3 IC50: 243 nM (HDAC1), 453 nM (HDAC2), 248 nM (HDAC3) Purity:98% Entinostat is the class I-selective HDAC inhibitor, with IC 50 of 243 nM, 453 nM, and 248 nM for HDAC1 , HDAC2 , and HDAC3 . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2541