Learn More
Selleck Chemical LLC Entospletinib
SDP
Supplier:  Selleck Chemical LLC S752310MG
Encompass_Preferred
Entospletinib (GS-9973) is an orally bioavailable selective Syk inhibitor with IC50 of 7 7 nM in a cell-free assay and showed 13- to 1000-fold cellular selectivity for Syk over other kinases(including Jak2 ckit Flt3 Ret KDR) as assessed by target protein phosphorylation or functional response
Catalog No. 50-000-00004