Learn More
TARGETMOL CHEMICALS INC Entrectinib 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T367825MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Entrectinib (RXDX-101) is a Trk, ROS1, and ALK inhibitor that inhibits TrkA, TrkB, TrkC, ROS1, and ALK (IC50=1/3/5/12/7 nM) with oral activity and blood-brain-barrier penetration. Entrectinib exhibits both antitumor and CNS activity. Purity 99.61%
Catalog No. 50-288-0080