Learn More
Medchemexpress LLC Entrectinib-d8 | 2251773-94-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12678S110MG
Encompass_Preferred
Entrectinib-d8 is a deuterated form of Entrectinib, an orally available inhibitor of TrkA/B/C, ROS1, and ALK receptors. It can cross the blood-brain barrier and demonstrates central nervous system activity. This compound is known to induce apoptosis and cell cycle arrest in cancer cells, showing anti-tumor activity and alleviation of bleomycin-induced pulmonary fibrosis in mice.
  • Deuterated compound
  • Orally available and blood-brain barrier permeable
  • Active in the central nervous system
  • Inhibits TrkA/B/C, ROS1, and ALK
  • Induces apoptosis and cycle arrest in cancer cells
  • Has anti-tumor activity
  • Alleviates bleomycin-induced pulmonary fibrosis
  • Useful as a tracer for drug development
  • May impact pharmacokinetic and metabolic profiles of drugs
  • Stable solid form
Catalog No. 50-003-41525