Learn More
Medchemexpress LLC envelope glycoprotei 50ug
SDP
Supplier:  Medchemexpress LLC HYP7412150UG
Encompass_Preferred
envelope glycoprotein gp120 Protein is a glycoprotein exposed on the surface of the HIV envelope Gp120 is essential for viral infection as it facilitates HIV entry into the host cell gp120 may also be facilitating viral persistence and continuing HIV infection by influencing the T cell immune response to the virus envelope glycoprotein gp120 Protein HIV-1 (AAA44191 HEK293 His) is the recombinant Virus-derived envelope glycoprotein gp120 protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-79092