Learn More
Medchemexpress LLC Envelope glycoprotein gp64 protein, AcmNPV (HEK293, His) | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7671110UG
Recombinant envelope glycoprotein gp64 from Autographa californica multiple nucleopolyhedrovirus (AcmNPV), expressed in HEK293 cells with a C-terminal His tag. The protein is supplied as a filtered PBS solution, with low endotoxin and characterized molecular weight, intended for biochemical and immunological applications. Store frozen and avoid repeated freeze-thaw cycles.
- Recombinant protein expressed in HEK293 cells.
- C-terminal His tag for detection and purification.
- Supplied as a 0.2 μm filtered PBS solution, pH 7.4.
- Endotoxin level <1 EU/μg as determined by LAL.
- Approximate molecular weight 54.2 kDa.
- Batch concentration and QC data available on the certificate of analysis.
- Store at -80°C; ship with dry ice; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.