Learn More
Medchemexpress LLC Envelope glycoprotein gp64 protein, AcmNPV (HEK293, His) | >95.0% | 100 UG

Supplier: Medchemexpress LLC HYP76711100UG
Recombinant envelope glycoprotein gp64 from Autographa californica multiple nucleopolyhedrovirus (AcmNPV), expressed in HEK293 cells and supplied as a C-terminal His-tagged protein. Supplied in PBS (0.2 μm filtered, pH 7.4) with low endotoxin and high purity, the protein is intended for biochemical and immunological research applications; store frozen and avoid repeated freeze-thaw cycles.
- Recombinant expression in HEK293 cells.
- C-terminal His tag for purification and detection.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Endotoxin level <1 EU/μg (LAL method).
- Supplied in 0.2 μm filtered PBS, pH 7.4.
- Concentration approximately 2.12 mg/mL (batch dependent).
- Stable at -80 °C; aliquot and avoid repeated freeze-thaw cycles.
- For research use only; not for human or clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.