Learn More
Waters Corp GPC Cleanup Column, 15 μm, 19 mm X 150 mm, 50 to 1K
Supplier: Waters Corp WAT036555
High-resolution GPC cleanup columns remove low volatility, high molecular weight interferences like lipids and natural resins from environmental samples, meeting EPA method 3640A. They enhance cleanup speed and reduce solvent consumption. Columns come packed in Methylene Chloride or Cyclohexane/Ethyl acetate to minimize solvent switchover.
- Removes low volatility, high molecular weight interferences like lipids and natural resins from environmental samples.
- Complies with EPA method 3640A.
- Increases cleanup speed and reduces solvent consumption.
- Features spherical polymer particles.
- pH range of 2 to 12.
- Available packed in Methylene Chloride or Cyclohexane/Ethyl acetate.
- Compatible with HPLC systems.
- Particle size: 15 μm.
- Inner diameter: 19 mm, Length: 150 mm.
- Molecular weight range: 50 to 1500.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.