Learn More
Medchemexpress LLC Envudeucitinibum | 2417135-66-9 | C22H18D6N6O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15370110MM/1ML
Encompass_Preferred
Envudeucitinibum is a highly selective, allosteric, and orally active TYK2 inhibitor that binds to the JH2 domain of TYK2. It exhibits no off-target effects on other kinases (JAK1-3). It reduces signaling and the production of proinflammatory cytokines including IL-12, IL-23, IL-17, and type I interferons (IFNs). It can be utilized for research related to plaque psoriasis, systemic lupus erythematosus (SLE), and other immune-mediated diseases.
  • Highly selective, allosteric, and orally active TYK2 inhibitor
  • Binds to the JH2 domain of TYK2
  • Exhibits no off-target effects on other kinases (JAK1-3)
  • Reduces signaling and production of proinflammatory cytokines
  • Potentially useful for research on immune-mediated diseases
Catalog No. 50-003-76011