Learn More
TARGETMOL CHEMICALS INC ENZALUTAMIDE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6002100MG
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 200 mg 500 mg 1000 mg 2000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Enzalutamide (MDV 3100) is a non-steroidal androgen receptor (AR) antagonist (IC50 36 nM in LNCaP prostate cells). purity: 99%
Catalog No. 50-257-9699