Learn More
Medchemexpress LLC HY-70002 10mg , Enzalutamide CAS:915087-33-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7000210MG
Encompass_Preferred
HY-70002 10mg Enzalutamide CAS: 915087-33-1 Enzalutamide (MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells. Enzalutamide is an autophagy activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9016