Learn More
Cayman Chemical EnzalutamIde 10mg
SDP
Supplier:  Cayman Chemical 1159610 MG
Encompass_Preferred
A non-steroidal androgen receptor antagonist (IC50 = 36 nM); reduces the efficiency of nuclear translocation of the androgen receptor and impairs both its binding to DNA and the recruitment of coactivators
Catalog No. 50-295-2954