Learn More
Selleck Chemical LLC Enzalutamide S1250-200mg
SDP
Supplier:  Selleck Chemical LLC S1250200MG
Encompass_Preferred
Enzalutamide is an androgen-receptor (AR) antagonist with IC50 of 36 nM in LNCaP cells Enzalutamide is shown to increase autophagy
Catalog No. 50-311-1214