Learn More
Medchemexpress LLC Enzalutamide | 915087-33-1 | MFCD14155804 | 99.97% | C21H16F4N4O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7000210MM/1ML
Encompass_Preferred
Enzalutamide, also known as MDV3100, is a potent androgen receptor (AR) antagonist that exhibits an IC50 of 36 nM in LNCaP prostate cells. It also functions as an autophagy activator. This product is for research use only.
  • Functions as an androgen receptor antagonist
  • Potent activity with IC50 of 36 nM in LNCaP prostate cells
  • Acts as an autophagy activator
  • Suitable for research applications
Catalog No. 50-004-33878